missing translation for 'onlineSavingsMsg'
Learn More

[(1H-Benzotriazol-1-yl)methyl]triphenylphosphonium Chloride 98.0+%, TCI America™

Catalog No. B44861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44861G 1 g
B44865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44861G Supplier TCI America Supplier No. B44861G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111198-09-5
Molecular Formula C25H21ClN3P
Molecular Weight (g/mol) 429.888
InChI Key YEEGDMCFIRPFRB-UHFFFAOYSA-M
PubChem CID 11988745
IUPAC Name benzotriazol-1-ylmethyl(triphenyl)phosphanium;chloride
SMILES C1=CC=C(C=C1)[P+](CN2C3=CC=CC=C3N=N2)(C4=CC=CC=C4)C5=CC=CC=C5.[Cl-]

Specifications

Melting Point 279°C
Color White
Quantity 1 g
Formula Weight 429.89
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material [(1H-Benzotriazol-1-yl)methyl]triphenylphosphonium Chloride