missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzimidazole-5-carbaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC45904DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC45904DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

58442-17-4
146.149
1h-benzimidazole-5-carbaldehyde, 1h-benzimidazole-5-carboxaldehyde, 1h-benzo d imidazole-5-carbaldehyde, benzimidazole-5-aldehyde, 1h-1,3-benzodiazole-5-carbaldehyde, 1h-benzimidazole-6-carboxaldehyde, benzimidazole-5-carboxaldehyde, 1h-benzoimidazole-5-carbaldehyde, 3h-benzoimidazole-5-carbaldehyde, pubchem9866
3H-benzimidazole-5-carbaldehyde
C8H6N2O
ALCHVVTYAHQOFY-UHFFFAOYSA-N
10012038
C1=CC2=C(C=C1C=O)NC=N2

Specifications

1H-Benzimidazole-5-carbaldehyde
97%
C8H6N2O
1h-benzimidazole-5-carbaldehyde, 1h-benzimidazole-5-carboxaldehyde, 1h-benzo d imidazole-5-carbaldehyde, benzimidazole-5-aldehyde, 1h-1,3-benzodiazole-5-carbaldehyde, 1h-benzimidazole-6-carboxaldehyde, benzimidazole-5-carboxaldehyde, 1h-benzoimidazole-5-carbaldehyde, 3h-benzoimidazole-5-carbaldehyde, pubchem9866
C1=CC2=C(C=C1C=O)NC=N2
146.149
146.15
58442-17-4
Amber Glass Bottle
5g
ALCHVVTYAHQOFY-UHFFFAOYSA-N
3H-benzimidazole-5-carbaldehyde
10012038
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents