missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzimidazole-5-carbaldehyde, 97%, Thermo Scientific™

Catalog No. p-4510683
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC45904CB 250mg Amber Glass Bottle
CC45904DA 1g Amber Glass Bottle
CC45904DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC45904CB Supplier Thermo Scientific Chemicals Supplier No. CC45904CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58442-17-4
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
InChI Key ALCHVVTYAHQOFY-UHFFFAOYSA-N
Synonym 1h-benzimidazole-5-carbaldehyde, 1h-benzimidazole-5-carboxaldehyde, 1h-benzo d imidazole-5-carbaldehyde, benzimidazole-5-aldehyde, 1h-1,3-benzodiazole-5-carbaldehyde, 1h-benzimidazole-6-carboxaldehyde, benzimidazole-5-carboxaldehyde, 1h-benzoimidazole-5-carbaldehyde, 3h-benzoimidazole-5-carbaldehyde, pubchem9866
PubChem CID 10012038
IUPAC Name 3H-benzimidazole-5-carbaldehyde
SMILES C1=CC2=C(C=C1C=O)NC=N2

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material 1H-Benzimidazole-5-carbaldehyde