missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Benzimidazole-2-carboxylic acid hydrate, 90%, Thermo Scientific™
Chemical Identifiers
| CAS | 849776-47-2 |
|---|---|
| Molecular Formula | C8H8N2O3 |
| Molecular Weight (g/mol) | 180.163 |
| InChI Key | YQHUUYLPOCNBKD-UHFFFAOYSA-N |
| Synonym | 1h-benzo d imidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid monohydrate, benzimidazole-2-carboxylic acid monohydrate, 1h-1,3-benzodiazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylicacid, hydrate 1:1, c8h6n2o2.h2o, 2-carboxy-1h-benzimidazole hydrate, 1h-benzimidazole-2-carboxylic acid-water 1/1 |
| PubChem CID | 18525703 |
| IUPAC Name | 1H-benzimidazole-2-carboxylic acid;hydrate |
| SMILES | C1=CC=C2C(=C1)NC(=N2)C(=O)O.O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
CC09051CB
|
N/A | 250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC09051DA
|
N/A | 1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC09051DE
|
N/A | 5g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 849776-47-2 | |
| 180.163 | |
| 1h-benzo d imidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid monohydrate, benzimidazole-2-carboxylic acid monohydrate, 1h-1,3-benzodiazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylicacid, hydrate 1:1, c8h6n2o2.h2o, 2-carboxy-1h-benzimidazole hydrate, 1h-benzimidazole-2-carboxylic acid-water 1/1 | |
| 1H-benzimidazole-2-carboxylic acid;hydrate |
| C8H8N2O3 | |
| YQHUUYLPOCNBKD-UHFFFAOYSA-N | |
| 18525703 | |
| C1=CC=C2C(=C1)NC(=N2)C(=O)O.O |
Specifications
| 849776-47-2 | |
| C8H8N2O3 | |
| YQHUUYLPOCNBKD-UHFFFAOYSA-N | |
| 1H-benzimidazole-2-carboxylic acid;hydrate | |
| 18525703 | |
| 90% |
| 90% | |
| 1h-benzo d imidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid monohydrate, benzimidazole-2-carboxylic acid monohydrate, 1h-1,3-benzodiazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylicacid, hydrate 1:1, c8h6n2o2.h2o, 2-carboxy-1h-benzimidazole hydrate, 1h-benzimidazole-2-carboxylic acid-water 1/1 | |
| C1=CC=C2C(=C1)NC(=N2)C(=O)O.O | |
| 180.163 | |
| 180.17 | |
| 1H-Benzimidazole-2-carboxylic acid hydrate |
Safety and Handling
Warning