missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzimidazole-2-carboxylic acid hydrate, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC09051DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC09051DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

849776-47-2
180.163
1h-benzo d imidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid monohydrate, benzimidazole-2-carboxylic acid monohydrate, 1h-1,3-benzodiazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylicacid, hydrate 1:1, c8h6n2o2.h2o, 2-carboxy-1h-benzimidazole hydrate, 1h-benzimidazole-2-carboxylic acid-water 1/1
1H-benzimidazole-2-carboxylic acid;hydrate
C8H8N2O3
YQHUUYLPOCNBKD-UHFFFAOYSA-N
18525703
C1=CC=C2C(=C1)NC(=N2)C(=O)O.O

Specifications

1H-Benzimidazole-2-carboxylic acid hydrate
90%
C8H8N2O3
1h-benzo d imidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylic acid monohydrate, benzimidazole-2-carboxylic acid monohydrate, 1h-1,3-benzodiazole-2-carboxylic acid hydrate, 1h-benzimidazole-2-carboxylicacid, hydrate 1:1, c8h6n2o2.h2o, 2-carboxy-1h-benzimidazole hydrate, 1h-benzimidazole-2-carboxylic acid-water 1/1
C1=CC=C2C(=C1)NC(=N2)C(=O)O.O
180.163
180.17
849776-47-2
Amber Glass Bottle
5g
YQHUUYLPOCNBKD-UHFFFAOYSA-N
1H-benzimidazole-2-carboxylic acid;hydrate
18525703
90%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents

Promotions

Promotions