missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,7H-Dodecafluoro-1-heptanol 97.0+%, TCI America™

Catalog No. p-7126596
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D110125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D110125G Supplier TCI America Supplier No. D110125G

Chemical Identifiers

CAS 335-99-9
Molecular Formula C7H4F12O
Molecular Weight (g/mol) 332.089
MDL Number MFCD00039630
InChI Key BYKNGMLDSIEFFG-UHFFFAOYSA-N
Synonym 1h,1h,7h-dodecafluoro-1-heptanol, 1,1,7-trihydroperfluoroheptanol, 1h,1h,7h-dodecafluoroheptanol, 1-heptanol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro, unii-lpx5k7ec3w, 1,1,7-trihydrododecafluoroheptanol, 1,1,7-trihydroperfluoroheptyl alcohol, 1-heptanol, 1h,1h,7h-dodecafluoro, 1,1,7-trihydrododecafluoroheptan-1-ol, 1h,1h,7h-dodecafluoro-1-hydroxyheptane
PubChem CID 67639
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptan-1-ol
SMILES C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 170°C
Quantity 25 g
Formula Weight 332.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,7H-Dodecafluoro-1-heptanol
RTECSNumber MJ4500000
TSCA Yes