accessibility menu, dialog, popup

UserName

HOFFFFFFFFFFFF

CAS RN 335-99-9

IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptan-1-ol
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptan-1-ol
Molecular Weight (g/mol): 332.09
Molecular Formula: C7H4F12O
InChi Key: BYKNGMLDSIEFFG-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)F
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)