missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,5H-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether 98.0+%, TCI America™
$93.21 - $296.38
Chemical Identifiers
| CAS | 16627-71-7 |
|---|---|
| Molecular Formula | C7H4F12O |
| Molecular Weight (g/mol) | 332.089 |
| MDL Number | MFCD00155904 |
| InChI Key | ZNBGTBKGFZMWKR-UHFFFAOYSA-N |
| Synonym | 2,2,3,3,4,4,5,5-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether |
| PubChem CID | 2782579 |
| IUPAC Name | 1,1,2,2,3,3,4,4-octafluoro-5-(1,1,2,2-tetrafluoroethoxy)pentane |
| SMILES | C(C(C(C(C(F)F)(F)F)(F)F)(F)F)OC(C(F)F)(F)F |
Chemical Identifiers
| 16627-71-7 | |
| 332.089 | |
| ZNBGTBKGFZMWKR-UHFFFAOYSA-N | |
| 2782579 | |
| C(C(C(C(C(F)F)(F)F)(F)F)(F)F)OC(C(F)F)(F)F |
| C7H4F12O | |
| MFCD00155904 | |
| 2,2,3,3,4,4,5,5-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether | |
| 1,1,2,2,3,3,4,4-octafluoro-5-(1,1,2,2-tetrafluoroethoxy)pentane |
Specifications
| 16627-71-7 | |
| 133°C | |
| MFCD00155904 | |
| 3271 | |
| ZNBGTBKGFZMWKR-UHFFFAOYSA-N | |
| 1,1,2,2,3,3,4,4-octafluoro-5-(1,1,2,2-tetrafluoroethoxy)pentane | |
| 2782579 | |
| ≥98.0% (GC) | |
| 1H,1H,5H-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether |
| Colorless | |
| C7H4F12O | |
| 5 g | |
| 2,2,3,3,4,4,5,5-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether | |
| C(C(C(C(C(F)F)(F)F)(F)F)(F)F)OC(C(F)F)(F)F | |
| 332.089 | |
| 332.09 | |
| Liquid |
Safety and Handling
TSCA : No