missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H-Perfluoro-1-hexene, 97%

Catalog No. p-7046929
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1683406 5 g
AAL1683414 25 g
AAL1683422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1683406 Supplier Thermo Scientific Chemicals Supplier No. L1683406

CAS: 19430-93-4 | C6H3F9 | 246.076 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19430-93-4
Molecular Formula C6H3F9
Molecular Weight (g/mol) 246.076
MDL Number MFCD00042338
InChI Key GVEUEBXMTMZVSD-UHFFFAOYSA-N
Synonym perfluorobutyl ethylene, 3,3,4,4,5,5,6,6,6-nonafluoro-1-hexene, perfluorobutylethylene, 1h,1h,2h-perfluoro-1-hexene, 1h,1h,2h-perfluorohexene, perfluorobutyl ethene, 1-hexene, 3,3,4,4,5,5,6,6,6-nonafluoro, 3,3,4,4,5,5,6,6,6-nonafluorohexene, perfluoro-n-butyl ethylene, 1h,1h,2h-perfluorohex-1-ene
PubChem CID 88054
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohex-1-ene
SMILES C=CC(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.457
Boiling Point 59°C to 60°C
Flash Point None
Refractive Index 1.289
Linear Formula CF3(CF2)3CH=CH2
Quantity 5 g
Beilstein 1819716
Formula Weight 246.08
Percent Purity 97%
Chemical Name or Material 1H,1H,2H-Perfluoro-1-hexene
Hazard Category H225
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
EINECSNumber 243-053-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only