missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane, 95%

Catalog No. p-7046862
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1660406 5 g
AAL1660414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1660406 Supplier Thermo Scientific Chemicals Supplier No. L1660406

CAS: 102488-47-1 | C10H10ClF13Si | 440.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102488-47-1
Molecular Formula C10H10ClF13Si
Molecular Weight (g/mol) 440.70
MDL Number MFCD00042359
InChI Key AKYGPHVLITVSJE-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorooctyldimethylchlorosilane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl dimethylchlorosilane, 2-tridecafluorohexyl ethyldimethylchlorosilane, tridecafluoro-1,1,2,2-tetrahydrooctyl-1-dimethylchlorosilane, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyldimethylchlorosilane, chloro-dimethyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-n-octyl silane, silane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl
PubChem CID 145382
IUPAC Name chloro-dimethyl-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
SMILES C[Si](C)(Cl)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Boiling Point 189°C to 191°C
Quantity 5 g
UN Number UN2986
Beilstein 7040835
Flash Point 52°C (125°F)
Formula Weight 440.71
Percent Purity 95%
Refractive Index 1.3453
Sensitivity Moisture sensitive
Density 1.461
Chemical Name or Material 1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, FLAMMABLE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only