accessibility menu, dialog, popup

UserName

CH3SiCH3ClFFFFFFFFFFFFF

CAS RN 102488-47-1

IUPAC Name: chlorodimethyl(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Synonyms: chlorodimethyl(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Molecular Weight (g/mol): 440.7
Molecular Formula: C10H10ClF13Si
InChi Key: AKYGPHVLITVSJE-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)