Learn More
1H,1H,2H,2H-Perfluorodecyltriethoxysilane, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L1658503
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications1H,1H,2H,2H-Perfluorodecyltriethoxysilane is used to bond poly(tetrafluoroethylene) films to silicon wafers. It is also utilized in anti-reflective coating, release coating and soil repellent coating.
Solubility
Miscible with ethanol and tetrahydrofuran.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 103°C to 106°C (3mmHg) | |
| 101947-16-4 | |
| C16H19F17O3Si | |
| MFCD00042334 | |
| 100°C (212°F) | |
| Miscible with ethanol and tetrahydrofuran. | |
| CCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC | |
| 610.38 | |
| 610.38 | |
| 1.342 | |
| 1.41 |
| 1H,1H,2H,2H-Perfluorodecyltriethoxysilane | |
| 1 g | |
| CF3(CF2)7(CH2)2Si(OCH2CH3)3 | |
| UN1760 | |
| 1h,1h,2h,2h-perfluorodecyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, heptadecafluoro-1,1,2,2-tetrahydrodecyl triethoxysilane, perfluorooctylethyl triethoxysilane, r-3-hydroxy-1-benyzl-pyrrolidine, triethoxy-1h,1h,2h,2h-perfluorodecylsilane, triethoxy-1h,1h,2h,2h-heptadecafluorodecylsilane, 1h,1h,2h,2h-perfluorodec-1-yl tris ethoxy silan, 1h,1h,2h,2h-perfluorodecyltriethoxysilane 1g | |
| MLXDKRSDUJLNAB-UHFFFAOYSA-N | |
| triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane | |
| 145374 | |
| 97% | |
| Moisture sensitive |
Chemical Identifiers
| 101947-16-4 | |
| 610.38 | |
| MLXDKRSDUJLNAB-UHFFFAOYSA-N | |
| 145374 | |
| CCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC |
| C16H19F17O3Si | |
| MFCD00042334 | |
| 1h,1h,2h,2h-perfluorodecyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, heptadecafluoro-1,1,2,2-tetrahydrodecyl triethoxysilane, perfluorooctylethyl triethoxysilane, r-3-hydroxy-1-benyzl-pyrrolidine, triethoxy-1h,1h,2h,2h-perfluorodecylsilane, triethoxy-1h,1h,2h,2h-heptadecafluorodecylsilane, 1h,1h,2h,2h-perfluorodec-1-yl tris ethoxy silan, 1h,1h,2h,2h-perfluorodecyltriethoxysilane 1g | |
| triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen