accessibility menu, dialog, popup

UserName

CH3OSiFFFFFFFFFFFFFFFFFOH3COH3C

CAS RN 101947-16-4

IUPAC Name: triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
Synonyms: perfluorooctylethyl triethoxysilane
Molecular Weight (g/mol): 610.38
Molecular Formula: C16H19F17O3Si
InChi Key: MLXDKRSDUJLNAB-UHFFFAOYSA-N
SMILES: CCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results