missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorodecyltriethoxysilane, 97%, Thermo Scientific™

Catalog No. AAL1658514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1658514 25 g
AAL1658503 1 g
AAL1658506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1658514 Supplier Thermo Scientific Chemicals Supplier No. L1658514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H,2H,2H-Perfluorodecyltriethoxysilane is used to bond poly(tetrafluoroethylene) films to silicon wafers. It is also utilized in anti-reflective coating, release coating and soil repellent coating.

Solubility
Miscible with ethanol and tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 101947-16-4
Molecular Formula C16H19F17O3Si
Molecular Weight (g/mol) 610.38
MDL Number MFCD00042334
InChI Key MLXDKRSDUJLNAB-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorodecyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, heptadecafluoro-1,1,2,2-tetrahydrodecyl triethoxysilane, perfluorooctylethyl triethoxysilane, r-3-hydroxy-1-benyzl-pyrrolidine, triethoxy-1h,1h,2h,2h-perfluorodecylsilane, triethoxy-1h,1h,2h,2h-heptadecafluorodecylsilane, 1h,1h,2h,2h-perfluorodec-1-yl tris ethoxy silan, 1h,1h,2h,2h-perfluorodecyltriethoxysilane 1g
PubChem CID 145374
IUPAC Name triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
SMILES CCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC

Specifications

Boiling Point 103°C to 106°C (3mmHg)
Quantity 25 g
Linear Formula CF3(CF2)7(CH2)2Si(OCH2CH3)3
UN Number UN1760
Flash Point 100°C (212°F)
Solubility Information Miscible with ethanol and tetrahydrofuran.
Formula Weight 610.38
Percent Purity 97%
Refractive Index 1.342
Sensitivity Moisture sensitive
Density 1.41
Chemical Name or Material 1H,1H,2H,2H-Perfluorodecyltriethoxysilane
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen