missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorodecyldimethylchlorosilane, tech. 90%, Thermo Scientific™

Catalog No. AAL1658206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1658206 5 g
AAL1658214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1658206 Supplier Thermo Scientific Chemicals Supplier No. L1658206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 74612-30-9
Molecular Formula C12H10ClF17Si
Molecular Weight (g/mol) 540.72
MDL Number MFCD00042343
InChI Key JHCJWHBMXWOYDE-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorodecyldimethylchlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl dimethylchlorosilane, 2-heptadecafluorooctyl ethylchlorodimethylsilane, chloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl dimethylsilane, chloro-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl-dimethylsilane, chlorodimethyl-3,3,4,4,5,5,6,6,7,7,8,8, 9,9,10,10,10-heptadecafluorodecylsilane, silane,chloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl dimethyl
PubChem CID 144665
IUPAC Name chloro-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)-dimethylsilane
SMILES C[Si](C)(CCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)Cl

Specifications

Boiling Point 197°C to 198°C
Quantity 5 g
UN Number UN2987
Beilstein 4584072
Formula Weight 540.73
Percent Purity 90%
Grade Technical
Refractive Index 1.3415
Sensitivity Moisture sensitive
Density 1.654
Chemical Name or Material 1H,1H,2H,2H-Perfluorodecyldimethylchlorosilane
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures