accessibility menu, dialog, popup

UserName

CAS RN 74612-30-9

IUPAC Name: chloro(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)dimethylsilane
Synonyms: chloro(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)dimethylsilane
Molecular Weight (g/mol): 540.72
Molecular Formula: C12H10ClF17Si
InChi Key: JHCJWHBMXWOYDE-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results