missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Nonafluorohexyl Acrylate (stabilized with TBC) 98.0+%, TCI America™

Catalog No. N11075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N11075G 5 g
N110725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N11075G Supplier TCI America Supplier No. N11075G

Chemical Identifiers

CAS 52591-27-2
Molecular Formula C9H7F9O2
Molecular Weight (g/mol) 318.14
MDL Number MFCD00236104
InChI Key GYUPEJSTJSFVRR-UHFFFAOYSA-N
Synonym Acrylic Acid 1H,1H,2H,2H-Nonafluorohexyl Ester, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Acrylate, Acrylic Acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Ester
PubChem CID 104247
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohexyl prop-2-enoate
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C

Specifications

Color Yellow
Quantity 5 g
UN Number 3272
Formula Weight 318.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,2H,2H-Nonafluorohexyl Acrylate (stabilized with TBC)
EINECSNumber (2)-4056
TSCA Yes