accessibility menu, dialog, popup

UserName

FFFFFFFFFOOH2C

CAS RN 52591-27-2

IUPAC Name: 3,3,4,4,5,5,6,6,6-nonafluorohexyl prop-2-enoate
Synonyms: 3,3,4,4,5,5,6,6,6-nonafluorohexyl prop-2-enoate
Molecular Weight (g/mol): 318.14
Molecular Formula: C9H7F9O2
InChi Key: GYUPEJSTJSFVRR-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results