missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H,3H,3H-Tridecafluoro-1-nonanol 98.0+%, TCI America™

Catalog No. p-7137661
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32585G 5 g
T325825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32585G Supplier TCI America Supplier No. T32585G
Only null left

Chemical Identifiers

CAS 80806-68-4
Molecular Formula C9H7F13O
Molecular Weight (g/mol) 378.133
MDL Number MFCD00077578
InChI Key HMGDEQANNRNNKX-UHFFFAOYSA-N
Synonym 3-(Perfluorohexyl)propanol
PubChem CID 2776181
IUPAC Name 4,4,5,5,6,6,7,7,8,8,9,9,9-tridecafluorononan-1-ol
SMILES C(CC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)CO

Specifications

Melting Point -3°C
Color Colorless
Boiling Point 86°C
Quantity 5 g
UN Number 1987
Formula Weight 378.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,2H,2H,3H,3H-Tridecafluoro-1-nonanol