Learn More
1H-1,2,4-Triazole-1-carboxamidine hydrochloride, 98%
CAS: 19503-26-5 | C3H6ClN5 | 147.566 g/mol
Supplier: Thermo Scientific Chemicals H6047506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1H-1,2,4-Triazole-1-carboxamidine hydrochloride | |
| 215°C to 220°C | |
| MFCD03095468 | |
| 9500271 | |
| JDDXNENZFOOLTP-UHFFFAOYSA-N | |
| 1,2,4-triazole-1-carboximidamide;hydrochloride | |
| 16218515 | |
| 98% |
| 19503-26-5 | |
| C3H6ClN5 | |
| 5 g | |
| 1h-1,2,4-triazole-1-carboximidamide hydrochloride, 1h-1,2,4-triazole-1-carboxamidine monohydrochloride, 1h-1,2,4-triazole-1-carboxamidine hydrochloride, 1,2,4-triazole-1-carboximidamide hydrochloride, 1h-1,2,4-triazole-1-carboximidamide, monohydrochloride, c3h5n5.clh, ksc496s5d, 1,2,4-triazolecarboxamidine, chloride, 1-amidino-1,2,4-triazole hydrochloride, 1-amidino-1h-1,2,4-triazole hydrochloride | |
| C1=NN(C=N1)C(=N)N.Cl | |
| 147.566 | |
| 147.57 |
Chemical Identifiers
| 19503-26-5 | |
| 147.566 | |
| JDDXNENZFOOLTP-UHFFFAOYSA-N | |
| 16218515 | |
| C1=NN(C=N1)C(=N)N.Cl |
| C3H6ClN5 | |
| MFCD03095468 | |
| 1h-1,2,4-triazole-1-carboximidamide hydrochloride, 1h-1,2,4-triazole-1-carboxamidine monohydrochloride, 1h-1,2,4-triazole-1-carboxamidine hydrochloride, 1,2,4-triazole-1-carboximidamide hydrochloride, 1h-1,2,4-triazole-1-carboximidamide, monohydrochloride, c3h5n5.clh, ksc496s5d, 1,2,4-triazolecarboxamidine, chloride, 1-amidino-1,2,4-triazole hydrochloride, 1-amidino-1h-1,2,4-triazole hydrochloride | |
| 1,2,4-triazole-1-carboximidamide;hydrochloride |
Safety and Handling
GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only