missing translation for 'onlineSavingsMsg'
Learn More

1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%

Selective inhibitor of nitric oxide-sensitive guanylyl cyclase | CAS: 41443-28-1 | C9H5N3O2 | 187.158 g/mol

Supplier:  Thermo Scientific Chemicals J62250MA

 View more versions of this product

Catalog No. AAJ62250MA

Only null left
Explore available promotions

Description

Description

Selective inhibitor of nitric oxide-sensitive guanylyl cyclase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one
160°C to 170°C
C9H5N3O2
10 mg
LZMHWZHOZLVYDL-UHFFFAOYSA-N
[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
1456
≥99%
41443-28-1
White
MFCD00792620
1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq
C1=CC=C2C(=C1)N=CC3=NOC(=O)N23
187.158
187.15
Solid

Chemical Identifiers

41443-28-1
187.158
LZMHWZHOZLVYDL-UHFFFAOYSA-N
1456
C1=CC=C2C(=C1)N=CC3=NOC(=O)N23
C9H5N3O2
MFCD00792620
1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq
[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : RO0913150

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only