missing translation for 'onlineSavingsMsg'
Learn More

1H-1,2,3-Triazolo[4,5-b]pyridine 98.0+%, TCI America™

Catalog No. T13671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13671G 1 g
T13675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13671G Supplier TCI America Supplier No. T13671G

Chemical Identifiers

CAS 273-34-7
Molecular Formula C5H4N4
Molecular Weight (g/mol) 120.12
MDL Number MFCD00134183
InChI Key VQNDBXJTIJKJPV-UHFFFAOYSA-N
Synonym 4-Azabenzotriazole
PubChem CID 136085
IUPAC Name 2H-[1,2,3]triazolo[4,5-b]pyridine
SMILES N1N=C2C=CC=NC2=N1

Specifications

Melting Point 207°C
Quantity 1 g
Formula Weight 120.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1H-1,2,3-Triazolo[4,5-b]pyridine