accessibility menu, dialog, popup

UserName

CAS RN 273-34-7

IUPAC Name: 2H-[1,2,3]triazolo[4,5-b]pyridine
Synonyms: 2H-[1,2,3]triazolo[4,5-b]pyridine
Molecular Weight (g/mol): 120.12
Molecular Formula: C5H4N4
InChi Key: VQNDBXJTIJKJPV-UHFFFAOYSA-N
SMILES: N1N=C2C=CC=NC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results