Learn More
1H-1,2,3-Triazole, 98%
CAS: 288-36-8 | C2H3N3 | 69.067 g/mol
Supplier: Thermo Scientific Chemicals A1699914
Description
It effectively promotes the proton conduction in polymer electrolyte membranes via an intermolecular proton-transfer mechanism. A useful triazole for proteomics research
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt effectively promotes the proton conduction in polymer electrolyte membranes via an intermolecular proton-transfer mechanism. A useful triazole for proteomics research
Solubility
Fully miscible in water.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications
| 1H-1,2,3-Triazole | |
| 23°C to 25°C | |
| 208°C | |
| C2H3N3 | |
| MFCD00014490 | |
| 104766 | |
| Fully miscible in water. | |
| C1=NNN=C1 | |
| 69.067 | |
| CHEBI:35565 | |
| 98% |
| 288-36-8 | |
| 1.192 | |
| 107°C (224°F) | |
| 1.498 | |
| 25 g | |
| 1h-1,2,3-triazole, 1,2,3-triazole, 2h-1,2,3-triazole, 1,2,3-1h-triazole, triazole, triazol, osotriazole, v-triazole, 1,2,3-triazol, 1,2,3 triazole | |
| QWENRTYMTSOGBR-UHFFFAOYSA-N | |
| 2H-triazole | |
| 67516 | |
| 69.07 |
Chemical Identifiers
| 288-36-8 | |
| 69.067 | |
| QWENRTYMTSOGBR-UHFFFAOYSA-N | |
| 67516 | |
| 2H-triazole |
| C2H3N3 | |
| MFCD00014490 | |
| 1h-1,2,3-triazole, 1,2,3-triazole, 2h-1,2,3-triazole, 1,2,3-1h-triazole, triazole, triazol, osotriazole, v-triazole, 1,2,3-triazol, 1,2,3 triazole | |
| CHEBI:35565 | |
| C1=NNN=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only