missing translation for 'onlineSavingsMsg'
Learn More

(1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetricarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7125265
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20635G 5 g
C206325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20635G Supplier TCI America Supplier No. C20635G
Only null left

Chemical Identifiers

CAS 16526-68-4
Molecular Formula C9H12O6
Molecular Weight (g/mol) 216.189
MDL Number MFCD00075294
InChI Key FTHDNRBKSLBLDA-UHFFFAOYSA-N
PubChem CID 349732
IUPAC Name cyclohexane-1,3,5-tricarboxylic acid
SMILES C1C(CC(CC1C(=O)O)C(=O)O)C(=O)O

Specifications

Melting Point 218°C
Color White-Yellow
Quantity 5 g
Formula Weight 216.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetricarboxylic Acid