accessibility menu, dialog, popup

UserName

CAS RN 16526-68-4

OHOOHOOOH

CAS RN 16526-68-4

IUPAC Name: cyclohexane-1,3,5-tricarboxylic acid
Synonyms: cyclohexane-1,3,5-tricarboxylic acid
Molecular Weight (g/mol): 216.19
Molecular Formula: C9H12O6
InChi Key: FTHDNRBKSLBLDA-UHFFFAOYSA-N
SMILES: OC(=O)C1CC(CC(C1)C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results