missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetricarboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 218°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 216.19 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetricarboxylic Acid |
Safety and Handling
| TSCA | No |