missing translation for 'onlineSavingsMsg'
Learn More

(1alpha,2alpha,4alpha)-1,2,4-Cyclohexanetricarboxylic Acid 98.0+%, TCI America™

Catalog No. C20625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20625G 5 g
C206225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20625G Supplier TCI America Supplier No. C20625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76784-95-7
Molecular Formula C9H12O6
Molecular Weight (g/mol) 216.189
InChI Key WTNDADANUZETTI-NGJCXOISSA-N
PubChem CID 12616203
IUPAC Name (1S,2R,4R)-cyclohexane-1,2,4-tricarboxylic acid
SMILES C1CC(C(CC1C(=O)O)C(=O)O)C(=O)O

Specifications

Melting Point 182°C
Color White
Quantity 5 g
Formula Weight 216.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1alpha,2alpha,4alpha)-1,2,4-Cyclohexanetricarboxylic Acid