accessibility menu, dialog, popup

UserName

OHOOOHOHO

CAS RN 76784-95-7

IUPAC Name: cyclohexane-1,2,4-tricarboxylic acid
Synonyms: cyclohexane-1,2,4-tricarboxylic acid
Molecular Weight (g/mol): 216.19
Molecular Formula: C9H12O6
InChi Key: WTNDADANUZETTI-UHFFFAOYNA-N
SMILES: OC(=O)C1CCC(C(C1)C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results