missing translation for 'onlineSavingsMsg'
Learn More

18-Bromo-1-octadecene 93.0+%, TCI America™

Catalog No. p-7119779
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B28161G 1 g
B28165G 5 g
2 options

Catalog No. B28161G

Supplier: TCI America B28161G

Only null left

Chemical Identifiers

CAS 172656-11-0
Molecular Formula C18H35Br
Molecular Weight (g/mol) 331.382
InChI Key HGBDROSSUKYLIG-UHFFFAOYSA-N
Synonym 17-Octadecenyl Bromide
PubChem CID 15192957
IUPAC Name 18-bromooctadec-1-ene
SMILES C=CCCCCCCCCCCCCCCCCBr

Specifications

Color Yellow
Quantity 1 g
Formula Weight 331.38
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 18-Bromo-1-octadecene
TSCA No
Recommended Storage Refrigerator