missing translation for 'onlineSavingsMsg'
Learn More

18-Bromo-1-octadecene 93.0+%, TCI America™

Numéro de catalogue. B28165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B28165G 5 g
B28161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B28165G Fournisseur TCI America Code fournisseur B28165G
Il en reste null

Identifiants chimiques

CAS 172656-11-0
Molecular Formula C18H35Br
Molecular Weight (g/mol) 331.382
InChI Key HGBDROSSUKYLIG-UHFFFAOYSA-N
Synonym 17-Octadecenyl Bromide
PubChem CID 15192957
IUPAC Name 18-bromooctadec-1-ene
SMILES C=CCCCCCCCCCCCCCCCCBr

Spécifications

Color Yellow
Quantity 5 g
Formula Weight 331.38
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 18-Bromo-1-octadecene
TSCA No
Recommended Storage Refrigerator