missing translation for 'onlineSavingsMsg'
Learn More
Learn More
18-Bromo-1-octadecene 93.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 331.38 |
| Percent Purity | ≥93.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 18-Bromo-1-octadecene |
Sécurité et manipulation
| TSCA | No |
| Recommended Storage | Refrigerator |