missing translation for 'onlineSavingsMsg'
Learn More

17alpha-Hydroxyprogesterone Acetate 98.0+%, TCI America™

Catalog No. H15271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15271G 1 g
H15275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15271G Supplier TCI America Supplier No. H15271G

Chemical Identifiers

CAS 302-23-8
Molecular Formula C23H32O4
Molecular Weight (g/mol) 372.51
MDL Number MFCD00021156
InChI Key VTHUYJIXSMGYOQ-UHFFFAOYNA-N
Synonym 17alpha-Acetoxyprogesterone, 17alpha-Hydroxypregn-4-ene-3,20-diene Acetate, 17alpha-Acetoxypregn-4-ene-3,20-dione
PubChem CID 11875276
IUPAC Name 1-acetyl-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
SMILES CC(=O)OC1(CCC2C3CCC4=CC(=O)CCC4(C)C3CCC12C)C(C)=O

Specifications

Melting Point 246°C
Color White-Yellow
Quantity 1 g
Formula Weight 372.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 17alpha-Hydroxyprogesterone Acetate
RTECSNumber TU5074000
TSCA No
Recommended Storage Refrigerator