accessibility menu, dialog, popup

UserName

CAS RN 302-23-8

IUPAC Name: 1-acetyl-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
Synonyms: hydroxyprogesterone acetate
Molecular Weight (g/mol): 372.51
Molecular Formula: C23H32O4
InChi Key: VTHUYJIXSMGYOQ-UHFFFAOYNA-N
SMILES: CC(=O)OC1(CCC2C3CCC4=CC(=O)CCC4(C)C3CCC12C)C(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results