missing translation for 'onlineSavingsMsg'
Learn More

17(H),21(H)-Hopane Solution, 0.1 mg/mL in isooctane, MilliporeSigma™ Supelco™

Catalog No. 111017950
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017950 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017950 Supplier Merck Emd Millipore Supplier No. 075621ML

17 β(H),21 β(H)-Hopanes belong to the hopanes family of hydrocarbons which are known as the biomarkers of petroleum products.

Featured Industries: Food and Beverages

Specifications

CAS 471-62-5
Flash Point -12°C
Linear Formula C30H52
MDL Number MFCD00216315
Synonym A′-Neogammacerane
Type Analytical Standard
Quantity 1 mL
Formula Weight 412.73
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 0.1 mg/mL in isooctane