missing translation for 'onlineSavingsMsg'
Learn More

16-Hydroxyhexadecanoic acid, 97%

Catalog No. p-7023864
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1784503 1 g
AAA1784506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1784503 Supplier Thermo Scientific Chemicals Supplier No. A1784503

CAS: 506-13-8 | C16H32O3 | 272.429 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 506-13-8
Molecular Formula C16H32O3
Molecular Weight (g/mol) 272.429
MDL Number MFCD00002750
InChI Key UGAGPNKCDRTDHP-UHFFFAOYSA-N
Synonym juniperic acid, 16-hydroxypalmitic acid, hexadecanoic acid, 16-hydroxy, palmitic acid, 16-hydroxy, omega-hydroxypalmitic acid, 16-hydroxy-hexadecanoic acid, unii-7ipp3u0f3i, .omega.-hydroxypalmitic acid, 7ipp3u0f3i, 16-hydroxy hexadecanoic acid
PubChem CID 10466
ChEBI CHEBI:55328
IUPAC Name 16-hydroxyhexadecanoic acid
SMILES C(CCCCCCCC(=O)O)CCCCCCCO

Specifications

Melting Point 98°C to 100°C
Linear Formula HO(CH2)15CO2H
Quantity 1 g
Beilstein 1783998
Formula Weight 272.43
Percent Purity 98%
Chemical Name or Material 16-Hydroxyhexadecanoic acid
EINECSNumber 208-028-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only