Learn More
Corning™ 1536-Well Nontreated Cyclic Olefin Copolymer Plates
GREENER_CHOICE
Nonsterile microplates have smooth top and clear, flat bottom.
$7,380.03 - $13,879.44
Specifications
Barcode Includes Barcode
Format Standard
Lid Without Lid
Array 32 x 48
Binding Type Not Treated
View More Specs
Products 2
Catalog Number Mfr. No. Color Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Color Quantity Price Quantity & Availability  
12456704
GREENER_CHOICE
View Documents Promotion Details
Corning™
4565
Black, Clear Bottom 10/Pk.
Case of 50 Each for $7,380.03
Only null left
12456700
View Documents Promotion Details
Corning™
4560
Black, Clear Bottom 20/Pk. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • 127 μm film thickness
  • 536-well low base, black/clear or white/clear bottom microplates
  • Bar coded
  • Custom bar codes available for compatibility with the Kalypsys system and with UHTS systems
  • Low auto-fluorescence
  • Broad chemical resistance including DMSO and alcohol
  • High mechanical stability
  • Optimized for flatness and uniformity
  • Low birefringence
Specifications
Specifications
Includes Barcode
Without Lid
Not Treated
Cycloolefin Copolymer
8 μL
Non-treated
For higher sensitivity
Standard
32 x 48
Black, Clear Bottom
1536
Non-sterile
Flat Bottom (F-well)