Learn More
Corning™ 1536-Well Nontreated Cyclic Olefin Copolymer Plates
Nonsterile microplates have smooth top and clear, flat bottom.
Supplier:  Corning™ 4560
 View more versions of this product
Catalog No. 12456700
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • 127 μm film thickness
  • 536-well low base, black/clear or white/clear bottom microplates
  • Bar coded
  • Custom bar codes available for compatibility with the Kalypsys system and with UHTS systems
  • Low auto-fluorescence
  • Broad chemical resistance including DMSO and alcohol
  • High mechanical stability
  • Optimized for flatness and uniformity
  • Low birefringence
Specifications
Chemical Identifiers
Includes Barcode
Without Lid
Not Treated
Cycloolefin Copolymer
8 μL
Non-treated
For higher sensitivity
Standard
32 x 48
Black, Clear Bottom
1536
Non-sterile
Flat Bottom (F-well)
Specifications
Includes Barcode
Without Lid
Not Treated
Cycloolefin Copolymer
8 μL
Non-treated
For higher sensitivity
20/Pk.
Standard
32 x 48
Black, Clear Bottom
1536
Non-sterile
Flat Bottom (F-well)
100/Cs.
20/Pk.
Product Suggestions
Product Suggestions