Learn More
Agilent BioTek 1536-well Microplate Wash Aspiration Manifold Kit
Catalog No. BT2120085
Change view
Click to view available options
For Use With (Equipment):
406 FX Washer Dispenser
Catalog No. For Use With (Equipment)
BT2120085 406 FX Washer Dispenser
1 options
Catalog No. BT2120085 Supplier Agilent BioTek Supplier No. 2120085
Microplate Wash Aspiration Manifold Kit for use with the 406 FX Washer Dispenser.
  • Includes aspiration manifold, alignment jig and instructions
  • Aspirate-only manifold is used with either a dual syringe pump dispenser module and 32-tube dispense manifold(s), or with a peristaltic pump dispenser module and a 1536-well microplate cassette
  • Dispense manifolds and cassettes are ordered separately
  • For syringe pump dispensing into 1536-well plates, at least one 32-tube dispense manifold is required
  • For faster dispensing, two 32-tube dispense manifolds can be used simultaneously
Specifications
Type Wash Module
For Use With (Equipment) 406 FX Washer Dispenser