Learn More
Greiner Bio-One 1536-well Low Base Polystyrene Microplates
For optical applications in high-content screening. Greiner Bio-One 1536 Well Low Base Polystyrene Microplates enables interference-free measurements and also microscopy over whole surface of bottom of microplate.
$6363.00 - $6363.00
Specifications
| Format | Standard |
|---|---|
| Array | 32 x 48 |
| Material | Polystyrene |
| No. of Wells | 1536 |
| Sterility | Non-sterile |
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Improved geometry
- Short distance of 0.3 mm between external well bottom and microplate skirt bottom
- Facilitates changing objectives during microscopy
Specifications
| Standard | |
| Polystyrene | |
| Non-sterile |
| 32 x 48 | |
| 1536 |