missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 1536-Well, Cell Culture-Treated, Flat-Bottom Microplate

Color: Black; Sterility: Sterile; Lid: With Lid

Supplier:  Corning™ 3726

 View more versions of this product

Catalog No. 07202512


Only null left

Description

Description

 

  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer FlexDrop™
  • Working volume of up to 10μL
  • Available untreated, with nonbinding surface (NBS™), or tissue culture treated
  • NBS microplates are ideal for homogeneous assays with “sticky” enzymes or for reducing nonspecific binding to microplate wells

 

Generic Barcode

  • Ensures compatibility with most bar code scanning and software systems
  • To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code (NT = not treated; TC = tissue culture treated; NB = nonbinding surface)
  • Labels applied to all four sides of the plate
  • No sequence number duplicates

 

Specifications

Chemical Identifiers

Standard
32 x 48
1536
Cell Culture
50/Cs.
With Lid
Black
Sterile
Flat Bottom (F-well)

Specifications

No Barcode
With Lid
32 x 48
Black
Polystyrene
8 μL
Cell Culture
Flat Bottom (F-well)
Black
Standard
0.03 cm2
10/Pk., 50/Cs.
1536-Well Assay Microplate
1536
Sterile
12.5 μL
50/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions