Learn More
Justrite™ 15 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Self-Closing Doors
GREENER_CHOICE
Catalog No. 19822327
Change view
Click to view available options
Configuration:
Left-Door
Right-Door
Color:
Light Neutral
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Configuration Color
19822327 Left-Door Light Neutral
19822332 Right-Door Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19822327 Supplier Justrite™ Supplier No. 882437
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 15 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Self-Closing Doors is fully compliant and designed for safe storage of solvents and flammables in laboratories.
  • Fill spouts have self-closing pressure-relief caps and stainless-steel flame arresters
  • Self-closing faucets provide trouble-free dispensing
Order Info
All cabinets measure D x W x H: 21.625 x 24 x 35.75 ft.
Specifications
Configuration Left-Door
Load Bearing Capacity 15 gal.
Door Style Self-close
Color Light Neutral