missing translation for 'onlineSavingsMsg'
Learn More

(15:0)-Anacardic acid, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013312
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013312 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013312 Supplier Merck Emd Millipore Supplier No. 0550610MG

(15:0)-Anacardic acid is a mixture of salicylic acid analogues, comprising of a 15 carbon aliphatic side chain in various degrees of unsaturation.

Featured Industries: Food and Beverages

Specifications

CAS 16611-84-0
Linear Formula C22H36O3
MDL Number MFCD07368254
Synonym Anacardic acid; 2-Hydroxy-6-pentadecylbenzoic acid; 22:0-Anacardic acid; 6-Pentadecylsalicylic acid
Type Analytical Standard
Quantity 10 mg
Formula Weight 348.52
Percent Purity ≥97% (HPLC)
Grade Analytical Standard