missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 14-Tooth Microplate Format Combs for Horizontal Midigel Systems

14-Tooth Microplate Format Combs for Horizontal Midigel Systems

$186.52 - $186.52

Specifications

For Use With (Equipment) Owl A1 Large Gel Electrophoresis System
Type Standard Comb
Format Microplate
Teeth Count 14
Width (English) Teeth 0.28 in.
View More Specs

Products 1
Catalog Number Mfr. No. Thickness (Metric) Teeth Volume (Fill) Metric Price Quantity  
Catalog Number Mfr. No. Thickness (Metric) Teeth Volume (Fill) Metric Price Quantity  
FB-SB-1334
View Documents
Fisherbrand™
A1MTC
0.1 cm 35 μL
Each for $186.52
Only null left
 
Description

Description

  • Accessory for use with Horizontal Midigel Systems

Specifications

Specifications

Owl A1 Large Gel Electrophoresis System
Microplate
0.28 in.
Standard Comb
14
0.72 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'