missing translation for 'onlineSavingsMsg'
Learn More

14-Heptacosanone, 97%

CAS: 542-50-7 | C27H54O | 394.728 g/mol

$188.89 - $693.77

Chemical Identifiers

CAS 542-50-7
Molecular Formula C27H54O
Molecular Weight (g/mol) 394.728
MDL Number MFCD00026598
InChI Key VCZMOZVQLARCOE-UHFFFAOYSA-N
Synonym 14-heptacosanone, ditridecyl ketone, myristone, unii-fq5vc928nn, fq5vc928nn, acmc-209leu, 14-heptacosanon, di-n-tridecyl ketone
PubChem CID 10955
IUPAC Name heptacosan-14-one
SMILES CCCCCCCCCCCCCC(=O)CCCCCCCCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2019106
View Documents
Thermo Scientific Chemicals
B2019106
5 g
Each for $188.89
Only null left
 
AAB2019114
View Documents
Thermo Scientific Chemicals
B2019114
25 g
Each for $693.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

542-50-7
394.728
VCZMOZVQLARCOE-UHFFFAOYSA-N
10955
CCCCCCCCCCCCCC(=O)CCCCCCCCCCCCC
C27H54O
MFCD00026598
14-heptacosanone, ditridecyl ketone, myristone, unii-fq5vc928nn, fq5vc928nn, acmc-209leu, 14-heptacosanon, di-n-tridecyl ketone
heptacosan-14-one

Specifications

542-50-7
C27H54O
5 g
14-heptacosanone, ditridecyl ketone, myristone, unii-fq5vc928nn, fq5vc928nn, acmc-209leu, 14-heptacosanon, di-n-tridecyl ketone
CCCCCCCCCCCCCC(=O)CCCCCCCCCCCCC
394.728
394.73
14-Heptacosanone
76°C to 79°C
MFCD00026598
1796794
VCZMOZVQLARCOE-UHFFFAOYSA-N
heptacosan-14-one
10955
97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-815-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only