missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 beta/zeta Monoclonal Antibody (22-IID8B), Invitrogen™
Description
Recommended positive controls: HeLa cell lysate. Store at 4oC short term. For extended storage aliquot and store at -20oC. Storage below -20oC is not recommended. Avoid freeze-thaw cycles.
14-3-3 beta/zeta Monoclonal antibody specifically detects 14-3-3 beta/zeta in Bovine, Canine, Human, Mouse, Non-human Primate, Ovine, Porcine, Rabbit, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.
Specifications
Specifications
| Antigen | 14-3-3 beta/zeta |
| Applications | Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 22-IID8B |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1mM PMSF, 50% glycerol and no preservative; pH 7.2 |
| Gene | YWHAB |
| Gene Accession No. | P35213, P63102, P63103, P68250, P31946, P63104, P63101, Q9CQV8 |
| Gene Alias | 1110013I11Rik; 1300003C17Rik; 14-3-3 alpha; 14-3-3 delta; 14-3-3 protein beta; 14-3-3 protein beta/alpha; 14-3-3 protein beta/alpha, N-terminally processed; 14-3-3 protein beta-subtype; 14-3-3 protein zeta; 14-3-3 protein zeta/delta; 14-3-3 protein/cytosolic phospholipase A2; 14-3-3 zeta; 14-3-3d; 14-3-3z; 14-3-3zeta; 14-3-3-zeta; 3-monooxygenase/tryptophan 5 monooxygenase activation protein beta polypeptide; AI596267; AL022924; AU020854; brain protein 14-3-3, beta isoform; epididymis luminal protein 4; epididymis secretory protein Li 1; epididymis secretory protein Li 3; epididymis secretory protein Li 93; factor activating exoenzyme S; FAS; GW128; HEL4; HEL-S-1; HEL-S-3; HEL-S-93; HS1; KCIP-1; mitochondrial import stimulation factor S1 subunit; Msfs1; phospholipase A2; prepronerve growth factor RNH-1; Protein 1054; Protein kinase C inhibitor protein 1; protein kinase C inhibitor protein-1; SEZ-2; tryosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta polypeptide; tryosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma; tyrosine 3/tryptophan 5 -monooxygenase activation protein beta polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, beta polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, zeta polypeptide; tyrosine 3/tryptophan 5-monooxygenase activation protein zeta polypeptide; tyrosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein beta polypeptide; tyrosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein, beta polypeptide; tyrosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein, beta polypeptide; tyrosine 3-monooxygenase; tyrosine 3-monooxygenase tryptophan 5-monooxygenase activation protein; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, alpha polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, delta polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta polypeptide; YWHAA; Ywhab; YWHAD; YWHAE; Ywhaz |
| Show More |