missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs 125 mL Wide Mouth Polypropylene Reagent Bottles with Polypropylene Caps
Description
- Bulk Pack of 500: 125 mL wide-mouth polypropylene reagent bottles with polypropylene caps, designed for safe liquid storage and lab applications.
- Autoclavable Construction: Autoclavable up to 135°C, making it suitable for sterilization and various lab environments requiring high temperature resistance.
- Wide Mouth Design: 28.8mm neck diameter offers easy filling, pouring, and handling of liquids, ensuring convenience in laboratory processes.
- Leakproof and Durable: Leak-proof tested for secure storage and transportation, with excellent chemical resistance for compatibility with diverse reagents.
- Translucent and Lightweight: Translucent material provides clear visibility of contents, and lightweight design ensures ease of handling and storage.
Specifications
Specifications
| Capacity (Metric) | 125 mL |
| Capacity (English) | 4.22 oz. |
| Color | Translucent |
| Diameter (Metric) Inner Neck | 28.8 mm |
| Material | Polypropylene |
| No. per Pack | 500 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Show More |