missing translation for 'onlineSavingsMsg'
Learn More

12-Tricosanone 95.0+%, TCI America™

Catalog No. T0404500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0404500G 500 g
T040425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0404500G Supplier TCI America Supplier No. T0404500G
Only null left

Chemical Identifiers

CAS 540-09-0
Molecular Formula C23H46O
Molecular Weight (g/mol) 338.62
MDL Number MFCD00026562
InChI Key VARQGBHBYZTYLJ-UHFFFAOYSA-N
Synonym 12-tricosanone, diundecyl ketone, lauron, laurone, undecyl ketone, di-n-undecyl ketone, lauron van, unii-5o6ejv7j5m, 5o6ejv7j5m, acmc-209lc3
PubChem CID 10888
IUPAC Name tricosan-12-one
SMILES CCCCCCCCCCCC(=O)CCCCCCCCCCC

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 500 g
Formula Weight 338.62
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 12-Tricosanone
EINECSNumber (2)-2475
RTECSNumber YD0805000
TSCA Yes