CAS RN 540-09-0
CAS RN 540-09-0
IUPAC Name:
tricosan-12-one
Synonyms:
12-tricosanone
Molecular Weight (g/mol):
338.62
Molecular Formula:
C23H46O
InChi Key:
VARQGBHBYZTYLJ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC(=O)CCCCCCCCCCC
1
–
1
of
1
results
1
12-Tricosanone 95.0+%, TCI America™
CAS: 540-09-0 Molecular Formula: C23H46O Molecular Weight (g/mol): 338.62 MDL Number: MFCD00026562 InChI Key: VARQGBHBYZTYLJ-UHFFFAOYSA-N Synonym: 12-tricosanone,diundecyl ketone,lauron,laurone,undecyl ketone,di-n-undecyl ketone,lauron van,unii-5o6ejv7j5m,5o6ejv7j5m,acmc-209lc3 PubChem CID: 10888 IUPAC Name: tricosan-12-one SMILES: CCCCCCCCCCCC(=O)CCCCCCCCCCC