missing translation for 'onlineSavingsMsg'
Learn More

12-Hydroxystearic acid, tech. 85%

CAS: 106-14-9 | C18H36O3 | 300.483 g/mol

Supplier:  Thermo Scientific Chemicals A1734722

 View more versions of this product

Catalog No. AAA1734722


Only null left

Description

Description

12-Hydroxystearic acid is used in cosmetics, wax blends, heavy duty greases, polishes, inks and hot melt adhesives, as a lubricant for natural and synthetic rubbers and as a source for industrial oleo chemicals. It is also used as an intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
12-Hydroxystearic acid is used in cosmetics, wax blends, heavy duty greases, polishes, inks and hot melt adhesives, as a lubricant for natural and synthetic rubbers and as a source for industrial oleo chemicals. It is also used as an intermediate in pharmaceuticals.

Solubility
Soluble in ether, alcohol and chloroform. Insoluble in water.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

12-Hydroxystearic acid
72°C to 77°C
CH3(CH2)5CH(OH)(CH2)10CO2H
100 g
12-hydroxystearic acid, octadecanoic acid, 12-hydroxy, harwax a, cerit fac 3, hydrofol acid 200, dl-12-hydroxystearic acid, ceroxin gl, barolub fto, loxiol g 21, stearic acid, 12-hydroxy
ULQISTXYYBZJSJ-UHFFFAOYSA-N
12-hydroxyoctadecanoic acid
7789
300.48
Technical
106-14-9
C18H36O3
MFCD00004592
1726730
Soluble in ether,alcohol and chloroform. Insoluble in water.
CCCCCCC(CCCCCCCCCCC(=O)O)O
300.483
CHEBI:85208
85%

Chemical Identifiers

106-14-9
300.483
ULQISTXYYBZJSJ-UHFFFAOYSA-N
7789
12-hydroxyoctadecanoic acid
C18H36O3
MFCD00004592
12-hydroxystearic acid, octadecanoic acid, 12-hydroxy, harwax a, cerit fac 3, hydrofol acid 200, dl-12-hydroxystearic acid, ceroxin gl, barolub fto, loxiol g 21, stearic acid, 12-hydroxy
CHEBI:85208
CCCCCCC(CCCCCCCCCCC(=O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-366-1

RTECSNumber : WI3700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only